tert-Butyl methyl ether
CHLCR00275820024CD
:
- Pack type:Glass bottle
- Pk:500 ml
- Seq:2725
:
:88.15 g/mol
:13,06059
:55.2 °C (1013 hPa)
:–108.6 °C
:0.7405 g/cm³ (20 °C)
:–28 °C
:Ambient
:216-653-1
:2398
:3,II
:GHS02
:GHS07
:1634-04-4
:(CH₃)₃COCH₃
:MFCD00008812