2-Nitroimidazole
Glass bottle for solids, 5 g
Catalog # CHLCR01338110144
Supplier: LOCAL DEALS
Jump to:
Product Details & Documents
Synonyms: Azomycin, 2-Nitro-1H-imidazole
- CAS Number:527-73-1
- Formula:C₃H₃N₃O₂
- MDL Number:MFCD00005185
- Molecular Weight:113.08 g/mol
- Merck Index:13,00923
- Melting Pt:279…284 °C
- Storage Temperature:Ambient
- EINECS:208-425-5
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



