L(+)-Ascorbic acid
Glass bottle for solids, 100 g
CHLCR02317020021
: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- :50-81-7
- :C₆H₈O₆
- :MFCD00064328
- :176.13 g/mol
- :13,00837
- :553 °C (1013 hPa)
- :190…192 °C
- :1.694 g/cm³ (20 °C)
- :Ambient
- :200-066-2
- :exempt
- Pack type:Glass bottle for solids
- Pk:100 g