订单输入
China
ContactUsLinkComponent
2-(1H-Pyrazol-5-yl)aniline ≥97.0% (通过GC)
刺激性
2-(1H-Pyrazol-5-yl)aniline ≥97.0% (通过GC)
Catalog # TCIAP1902-1G
供应商:  TCI
CAS 编号:  
undefined
刺激性
2-(1H-Pyrazol-5-yl)aniline ≥97.0% (通过GC)
Catalog # TCIAP1902-1G
供应商:  TCI
CAS 编号:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

规格

  • 包装类型:
    Glass bottle
  • 包装规格:
    1 g
  • 顺序:
    3047

规格

规格测试结果

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Nonaqueous Titration):
    min. 95.0 %

规格测试结果

关于此项目

 :
 :159.1879 g/mol
熔点:129°C
 :GHS07
CAS 编号:111562-32-4
化学式:C₉H₉N₃
MDL 编号:MFCD09028476