Order Entry
China
ContactUsLinkComponent
Tetrabutylammonium hydroxide 10% in aqueous solution
Corrosive
Tetrabutylammonium hydroxide 10% in aqueous solution
Catalog # CHLCR00920720134
Supplier:  LOCAL DEALS
CAS Number:  
Corrosive
Tetrabutylammonium hydroxide 10% in aqueous solution
Catalog # CHLCR00920720134
Supplier:  LOCAL DEALS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :259.48 g/mol
Storage Temperature:Refrigerator
EINECS:218-147-6
UN:3267
ADR:8,II
 :GHS05
CAS Number:2052-49-5
Formula:C₁₆H₃₇NO
MDL Number:MFCD00009425