1H-Pyrazole-3-carboxylic acid
Catalog # CHLCR01838410044
Supplier: LOCAL DEALS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:112.09 g/mol
CAS Number:1621-91-6
Formula:C₄H₄N₂O₂
MDL Number:MFCD00077436