Order Entry
China
ContactUsLinkComponent
O-Dianisidine dihydrochloride ≥98.0% (by HPLC) for biochemical research
Irritant
Health
O-Dianisidine dihydrochloride ≥98.0% (by HPLC) for biochemical research
Supplier:  TCI
Certificates
undefined
Irritant
Health
O-Dianisidine dihydrochloride ≥98.0% (by HPLC) for biochemical research
Supplier:  TCI

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Appearance:
    White to Gray to Red powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 95.0 %
  • Peroxidase detection test:
    to pass test

Specification Test Results

Related Information