Order Entry
China
ContactUsLinkComponent
Sodium-4-aminonaphthalene-1-sulphonate tetrahydrate ≥95.0% (by total nitrogen basis)
Irritant
Sodium-4-aminonaphthalene-1-sulphonate tetrahydrate ≥95.0% (by total nitrogen basis)
Catalog # TCIAA0353-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Sodium-4-aminonaphthalene-1-sulphonate tetrahydrate ≥95.0% (by total nitrogen basis)
Catalog # TCIAA0353-25G
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (with Total Nitrogen):
    min. 95.0 % (calcd.on anh.substance)
  • Water:
    20.0 to 25.0 %

Specification Test Results

About this item

 :
 :245.23 g/mol
Merck Index:14,06403
Melting Pt:280 °C
EINECS:204-975-5
 :GHS07
CAS Number:130-13-2
Formula:C₁₀H₈NNaO₃S.4H₂O
MDL Number:MFCD00044844