Order Entry
China
ContactUsLinkComponent
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride ≥98.0% (by HPLC) for biochemical research
Irritant
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride ≥98.0% (by HPLC) for biochemical research
Supplier:  TCI
Certificates
undefined
Irritant
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride ≥98.0% (by HPLC) for biochemical research
Supplier:  TCI

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 97.0 % (calcd.on dried substance)
  • Drying loss:
    max. 6.0 %
  • Alkaline Phosphatase detection test:
    to pass test v

Specification Test Results

Related Information