Order Entry
China
ContactUsLinkComponent
Ion exchange columns, ZORBAX® SAX and SCX
Ion exchange columns, ZORBAX® SAX and SCX
  HEWL860700-704
Ion exchange columns, ZORBAX® SAX and SCX
  HEWL860700-704

 

  • Column Format:
    Analytical
  • Ø int.:
    2,10 mm
  • Particle Size:
    5 µm
  • Pore size:
    300 Å
  • Max. pressure:
    350 bar
  • Description:
    Narrow bore
  • Length:
    50 mm
  • Seq:
    09
  • USP code:
    L50
  • pH range:
    2 - 7
  • Surface area:
    50 m²/g
  • Phase:
    SCX
  • Pk:
    1

 

 

Both strong anion exchange (SAX) and strong cation exchange (SCX) columns are bonded with spherical silica particles for optimum efficiency. SAX is ideal for separation of water-soluble compounds such as aromatic and aliphatic carboxylic acids and sulphonic acids and SCX is ideal for separations of basic, water-soluble compounds and bio-molecules.

  • Stable from pH 2 to 7
  • Provide high efficiency, rapid separations
  • Compatible with organic mobile phase modifiers
  • A trifunctional organo silane reagent is used in producing ZORBAX® SAX packing to maximise its stability with aqueous mobile phases