Pipette tips, racked, for Pos-D™ pipettes
Supplier: RAININ
Certificates
About this item
These tips are designed for Pos-D™ positive displacement pipettes. A disposable piston, which moves within a plastic capillary, makes direct contact with the liquid. The positive wiping action of the piston against the capillary wall assures complete dispensing without droplets. These tips eliminate cross-contamination from aerosols, samples or pipette.