Bio SEC-3 columns are packed with spherical, narrowly dispersed 3 µm silica particles coated with a proprietary hydrophilic layer for high recovery and minimal secondary interactions which provides more consistent separations.
- Compatible with most aqueous buffers
- Stable under high and low salt condition
Bio SEC- 5 columns are packed with 5 µm silica particles coated with a proprietary, neutral, hydrophilic layer for maximum efficiency and stability.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- HEWL5190-2534
- HEWL5190-2503
- HEWL5190-2539
- Max. pressure:
- 2000 psi
- 2000 psi
- 2000 psi
- pH range:
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- Temperature limit:
- 10...20 °C
- 10...20 °C
- 10...20 °C