Order Entry
China
Orders LinkContactUsLinkComponent
VWR-Ecotek-540x390.png
EcoTek Sustainable Technology
Available in many popular PowerForm®, TrueForm®, and MegaMan® single-use styles, EcoTek® is a proprietary nitrile treatment that accelerates waste reduction in landfills. SW® nitrile gloves with EcoTek reduce waste by 92.6% in only 2 years and 6 months under test method ASTM D5526-12, a standard test method for determining anaerobic biodegradation of plastics in accelerated landfill conditions. Current biodegradation results do not predict future results.
EcoTek Sustainable Gloves | Avantor
Product selection
PF-065-095-NRG-ECO-BAL-GR-WT_Thumbnail.jpg
Experience EnerGel infused gloves, designed to hydrate skin and protect against long-term glove usage.
PF-088-095-DRKECO-GY_Thumbnail.jpg
Work with confidence and comfort in environmentally conscious sustainable gloves with our exclusive DriTek sweat management technology.
PF-065-090-ECO-BK_Thumbnail.jpg
Protect hands—and the environment—with a sustainable single-use glove that combines performance, durability and tactile sensitivity for critical tasks.
PF-065-095-ECO-TL_Thumbnail.jpg
Versatile sustainable 5 mil single-use glove combines maximum comfort and control with high-quality construction and high-performance protection.