Order Entry
China
ContactUsLinkComponent
Ultrapure-Water-Systems.jpg
Ultrapure Water Systems
Our Arium® laboratory-grade water purification systems feature an inspiring, application-oriented design. They enable you to perform your workflows faster and more reliably, simplifying your daily lab work while ensuring cost-efficient operation over the long term. 
vwr_child_page_Lab_Water_arium_mini.jpg
Ultrapure water system ideal for daily comsumption up to 10 liters of water per day
vwr_child_page_Lab_Water_arium_pro.jpg
Arium® Pro Ultrapure Water Systems
Ultrapure water systems that meet or exceed ASTM Type I water for any application
vwr_child_page_Lab_Water_arium_comfort.jpg
Arium® Comfort All in One Water Systems
The Arium® Comfort Systems provide Type 1 and Type 3 or Type 1 and Type 2 water in a combined system.
vwr_child_page_Lab_Water_arium_smart_station.jpg
The Arium® Smart Station Ultrapure is designed for flexible remote dispensing of ultrapure water from Arium® Pro and Arium® Comfort lab water systems, directly at the point of use.