Order Entry
China
Orders LinkContactUsLinkComponent
BucketsCartsAndWringersForEfficientCleaning-955x690.jpg
Buckets, Carts, and Wringers for Efficient Cleanin
Critical cleaning relies heavily on the use of correct tools. Micronova’s variety of carts, multiple bucket configurations and wringers ensures proper disinfectant and detergent application while allowing for increased clean rinse options.
Buckets, Carts, and Wringers for Efficient Cleaning | Avantor
BucketBinderMultiBucketSystems-400x495.jpg
The BucketBinder Multi Bucket System is a cost efficient, ergonomic, and simple multi bucket system that does away with the need for bulky carts and frames. The system easily disassembles and offers easier cleaning and storage – in both small and large sizes.
SlimTBuckets-400x495.jpg
Rectangular buckets with a 19" interior width for use with a variety of flat head mops and string mops in controlled environments - with and without casters. Buckets are autoclavable.
SlimTDoubleBucketCart-400x495.jpg
A low profile, electropolished stainless steel frame that secures and easily transports cleaning buckets (included) with excellent maneuverability. Buckets available in stainless steel or polypropylene.
SlimLineStainlessSteelCartSystems-400x495.jpg
A durable cart system crafted from electropolished stainless steel for it's frame, optional wringer bar, and three grip-it holders for easy storage and interchange of handles and mop heads. Available as a double or triple bucket system.