Order Entry
China
Orders LinkContactUsLinkComponent
654926_AMEA-BPP-CEC-campaign_page-header_CR_web_955x590px_EN_v1.jpg
Power Your Process. Protect What Matters.
We manage your cleanroom protection so you can focus on innovation, compliance and faster delivery of high-quality therapies to patients.
Power Your Process. Protect What Matters. | Avantor
From sterile apparel to disinfectants and wipes, Avantor® offers validated, high-performance solutions to help you maintain cleanroom integrity. Our comprehensive portfolio supports contamination control, compliance and operational efficiency across every stage of biopharmaceutical production. With trusted products and expert guidance behind you, you can focus on advancing science, accelerating timelines and delivering high-quality therapies with confidence.
Equip Your Process with Cleanroom Essentials
content_block__img01.jpg
Find the Right Fit for Your Cleanroom
Explore cleanroom consumables by application area: gowning, cleaning, monitoring or packaging and get the exact fit for your workflow.
content_block__img02.jpg
Solutions for Your Industry
We understand your standards. Find curated contamination control products and support for biopharma, medical device, electronics, aerospace and beyond.
content_block__img03.jpg
Brands You Trust, Quality You Need
Discover proven cleanroom brands: validated, documented and ready for inspection. Only the suppliers that meet your standards make the list.
content_block__img04.jpg
Sustainable Choices for Your Cleanroom
Choose cleanroom products designed to reduce your environmental impact: sustainably produced, thoughtfully packaged or fully recyclable, without compromising safety or compliance.
content_block__img05.jpg
Services That Support Your Process
Consulting, documentation and more. See how our expert services can simplify your cleanroom compliance and operational goals.
Sustainable-Solution.jpg
Sustainable Solutions
Review our more ecofriendly safety and cleanroom supplies – either sustainability produced and packaged or recycled.
Understanding Cleanrooms and Controlled Environments
What is a Controlled Environment?
A controlled environment is one in which multiple environmental factors are monitored and controlled using controlled environment equipment. The controlled parameters typically include:
  • Particulates
  • Humidity
  • Temperature
  • Pressure
  • Segregation
What is a Cleanroom?
Cleanrooms are controlled environments that limit the number of pollutants, or contamination, to a specified number of particles (by size) per cubic foot or cubic meter. Cleanrooms are tested regularly to ensure standards are met and systems are in place to control the environment by limiting dust, chemical vapors, airborne microbes and aerosol particles.
Cleanrooms are used by manufacturing companies when a high level of sterility is necessary to avoid adversely affecting the product being manufactured. Cleanroom solutions vary based on the manufacturer’s needs and are used in a variety of industries, including:
  • Pharmaceutical
  • Biotech
  • Medical device
  • Life sciences
  • Aerospace
  • Semiconductor manufacturing
  • Optics
  • Military
  • Department of Energy
The Standards for a Cleanroom
The standards for a cleanroom are based on the number of particles measured in one cubic foot or one cubic meter of air. Cleanrooms are classified using either the Federal Standard 209E of the USA or ISO 14644-1 standard.
Federal Standard 209E classifies cleanrooms with numbers like “class 1,000” or “class 10,000,” and classifications are based on the number of particles of size 0.5 μm or larger allowed in one cubic foot of air.
ISO 14644-1 standard classifies cleanrooms with smaller numbers like “3” and classifications are based on a decimal logarithm of the number of particles 0.1 μm or larger allowed in one cubic meter of air.
What Lab Equipment and Supplies You Need
The equipment and supplies required for a controlled environment depend on the cleanroom’s classification and compliance standards. Common cleanroom equipment and supplies include:
  • High-Efficiency Particulate Air (HEPA) filters
  • Ultra-Low Particulate Air (ULPA) filters
  • Airlocks
  • Air showers
  • Special gowning designed to trap contaminants
  • Environmental monitoring
Cleanroom 101 Resource Center
res_center--apparel.jpg
Find validated cleanroom garments and gloves tailored for your industry, standards and workflow needs.
EXPLORE NOW
res_center--workflow_planning.jpg
See how our solutions map to your cleanroom, from entry to fill and
finish. They are ideal for validation and procurement planning.
READ NOW
res_center--resources.jpg
Explore a curated collection of cleanroom literature. Find everything you need to understand classifications, contamination control, best practices and validation support.
EXPLORE
res_center--gloves.jpg
Support your cleanroom processes with sterile and nonsterile nitrile gloves that deliver tactile sensitivity, chemical resistance and low particle counts. Tested to ASTM, FTMS, and IEST standards.
LEARN MORE
flyer-cleanroom.jpg
Insights and expert interviews on contamination control, sustainability and regulatory trends in biopharma manufacturing.
DOWNLOAD NOW
Focus on Innovation. Let Us Handle the Cleanroom.
Get expert support and trusted solutions that protect your cleanroom,
so you can focus on what drives science forward.
CONTACT US