ContactUsLinkComponent
70% alcohol processed with deionized water for cleanroom use, 0.2 micron filtered, and simply packaged 1 * 6 items
Codice catalogo SHIE6600605
Nome del fornitore:  ECOLAB CONTAMINATION CONTROL
New Product
70% alcohol processed with deionized water for cleanroom use, 0.2 micron filtered, and simply packaged 1 * 6 items
Codice catalogo SHIE6600605
Nome del fornitore:  ECOLAB CONTAMINATION CONTROL