Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Human Recombinant MARCH2 (from E. coli)

Human Recombinant MARCH2 (from E. coli)

Codice catalogo ABCAAB314524-100UG
New Product
Nome del fornitore:   Abcam
Human Recombinant MARCH2 (from E. coli)
Codice catalogo ABCAAB314524-100UG
Nome del fornitore:  Abcam
New Product
Numero CAS:  
Human Recombinant MARCH2 (from E. coli)
Human Recombinant MARCH2 (from E. coli)
Numero CAS:  
Price
3 820.00 SFr.
Log in per disponibilità
1 * 100 µG
Product Details & Documents

Recombinant human MARCH2 protein (His tag) is a Human Full Length protein, in the 1 to 246 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifiche

Specifiche

  • Senza animali:
    Y
  • Protein/peptide name:
    MARCH2
  • Purezza:
    <gt/>90%
  • Attività biologica:
    Standard
  • Protein synonyms:
    MARCH2|RNF172|HSPC240|MARCHF2|E3 ubiquitin-protein ligase MARCHF2|Membrane-associated RING finger protein 2|Membrane-associated RING-CH protein II|RING finger protein 172|RING-type E3 ubiquitin transferase MARCHF2|MARCH-II
  • Specie:
    Human
  • Coniugazione:
    Non coniugato
  • Sorgente luminosa:
    E. coli
  • Cat. No.:
    ABCAAB314524-100UG
  • Condizioni di conservazione:
    Store at −20 °C
  • Protein/peptide type:
    Recombinant
  • Forma:
    Lyophilised
  • Applicazioni testate:
    SDS-PAGE
  • Formulazione:
    pH: 7,4 - 8, Constituents: 6% Trehalose, 0,14% PBS
  • Temperatura di spedizione:
    20

Specifiche