Orders LinkContactUsLinkComponent
CentriVap™ Cold Traps
CentriVap™ Cold Traps
CentriVap cold trap −50 °C with acrylic lid, 230 V, 50 Hz, Schuko, 40,9×61,5×35,1 cm
Codice catalogo LBCP8601030
New Product
Nome del fornitore:   LABCONCO
CentriVap™ Cold Traps
Codice catalogo LBCP8601030
Nome del fornitore:  LABCONCO
New Product
CentriVap™ Cold Traps
CentriVap™ Cold Traps
CentriVap cold trap −50 °C with acrylic lid, 230 V, 50 Hz, Schuko, 40,9×61,5×35,1 cm
Price
SFr. 5’980.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
CentriVap™ cold traps collect evaporated liquid from a CentriVap concentrator, protecting the vacuum pump and laboratory environment from vapors and fumes.
  • Brushed, stainless steel and powder-coated steel exterior
  • Illuminated temperature graph with endpoint indication
  • HCFC-free refrigerant system
  • 4-litre, stainless steel collector
  • Acrylic or stainless steel lid with two Quick-Disconnect fittings for connection to a CentriVap concentrator
  • Stainless steel drain port (with corrosion-resistant, nylon valve)
  • −50 °C models feature a ¹/₃ hp compressor (−50 °C in less than 30 min)
  • −84 °C models feature two ¹/₃ hp compressors (−84 °C in less than 40 min)
  • −105 °C models feature two ¹/₃ hp compressors (−105 °C in less than 40 min)
Certifications: UL, CAN/CSA C22.2.
Specifiche
  • Descrizione:
    CentriVap cold trap −50 °C with acrylic lid, 230 V, 50 Hz, Schuko, 40,9×61,5×35,1 cm
  • Seq:
    4