ContactUsLinkComponent
1 component: Nickel (Ni 2+) 10mg/l in Water CRM, ISO 17034 and ISO 17025,,,, conservation 12 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C,,,, PACKAGING 100 ML BOTTLE 1 * 100 mL
Catalog # CMPLRM120888L1
New Product
1 component: Nickel (Ni 2+) 10mg/l in Water CRM, ISO 17034 and ISO 17025,,,, conservation 12 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C,,,, PACKAGING 100 ML BOTTLE 1 * 100 mL
Catalog # CMPLRM120888L1