ContactUsLinkComponent
Monkey IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antib ody, Streptavidin-HRP, recombinant human IL-17A ELISA standard, and standard reconstitution buffer. This matched anti 1 * 576 Tests
Codice catalogo ANTIA270511-696
Nome del fornitore:  ANTIBODIES.COM
New Product
Monkey IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antib ody, Streptavidin-HRP, recombinant human IL-17A ELISA standard, and standard reconstitution buffer. This matched anti 1 * 576 Tests
Codice catalogo ANTIA270511-696
Nome del fornitore:  ANTIBODIES.COM