Mouse IL-6 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-AP, recombinant mouse IL-6 ELISA standard, and standard reconstitution buffer. This matched antibody p 1 * 1.920 Tests
Codice catalogo ANTIA270446-2096
Nome del fornitore: ANTIBODIES.COM