ContactUsLinkComponent
Human IL-1 alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-AP, recombinant human IL-1a ELISA standard, and standard reconstitution buffer. This matched ant 1 * 576 Tests
  ANTIA270398-696
New Product
Human IL-1 alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-AP, recombinant human IL-1a ELISA standard, and standard reconstitution buffer. This matched ant 1 * 576 Tests
  ANTIA270398-696