Llama Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detect ion antibody, Streptavidin-AP, recombinant llama Interferon gamma ELISA standard, and standard reconstitution buffer. 1 * 576 Tests
Codice catalogo ANTIA270422-696
Nome del fornitore: ANTIBODIES.COM