Bovine IL-2 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-AP, recombinant bovine IL-2 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 576 Tests
Codice catalogo ANTIA270375-696
Nome del fornitore: ANTIBODIES.COM