ContactUsLinkComponent
Llama Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detect ion antibody, Streptavidin-HRP, recombinant llama Interferon gamma ELISA standard, and standard reconstitution buffer 1 * 1.920 Tests
Codice catalogo ANTIA270506-2096
Nome del fornitore:  ANTIBODIES.COM
New Product
Llama Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detect ion antibody, Streptavidin-HRP, recombinant llama Interferon gamma ELISA standard, and standard reconstitution buffer 1 * 1.920 Tests
Codice catalogo ANTIA270506-2096
Nome del fornitore:  ANTIBODIES.COM