ContactUsLinkComponent
Human IL-10 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant human IL-10 ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 1.920 Tests
Codice catalogo ANTIA270484-2096
Nome del fornitore:  ANTIBODIES.COM
New Product
Human IL-10 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant human IL-10 ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 1.920 Tests
Codice catalogo ANTIA270484-2096
Nome del fornitore:  ANTIBODIES.COM