ContactUsLinkComponent
Human IgG Matched Antibody Pair Kit (High Sensitivity) includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-HRP, native human IgG ELISA standard, and standard reconstitution buffer. This matche 1 * 576 Tests
Catalog # ANTIA270481-696
Supplier:  ANTIBODIES.COM
New Product
Human IgG Matched Antibody Pair Kit (High Sensitivity) includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-HRP, native human IgG ELISA standard, and standard reconstitution buffer. This matche 1 * 576 Tests
Catalog # ANTIA270481-696
Supplier:  ANTIBODIES.COM