Mouse IgM Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-HRP, Purified mouse IgM ELISA standard, and standard reconstitution buffer A5. This matched antibody pa 1 * 576 Tests
Catalog # ANTIA270518-696
Supplier: ANTIBODIES.COM