ContactUsLinkComponent
Chicken Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal dete ction antibody, Streptavidin-AP, recombinant chicken Interferon gamma ELISA standard, and standard reconstitution buf 1 * 576 Tests
Codice catalogo ANTIA270378-696
Nome del fornitore:  ANTIBODIES.COM
New Product
Chicken Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal dete ction antibody, Streptavidin-AP, recombinant chicken Interferon gamma ELISA standard, and standard reconstitution buf 1 * 576 Tests
Codice catalogo ANTIA270378-696
Nome del fornitore:  ANTIBODIES.COM