ContactUsLinkComponent
Human IL-23 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant human IL-23 ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 576 Tests
Codice catalogo ANTIA270492-696
Nome del fornitore:  ANTIBODIES.COM
New Product
Human IL-23 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant human IL-23 ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 576 Tests
Codice catalogo ANTIA270492-696
Nome del fornitore:  ANTIBODIES.COM