ContactUsLinkComponent
Human Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detect ion antibody, Streptavidin-HRP, recombinant human Interferon gamma ELISA standard, and standard reconstitution buffer 1 * 1.920 Tests
Catalog # ANTIA270479-2096
Supplier:  ANTIBODIES.COM
New Product
Human Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detect ion antibody, Streptavidin-HRP, recombinant human Interferon gamma ELISA standard, and standard reconstitution buffer 1 * 1.920 Tests
Catalog # ANTIA270479-2096
Supplier:  ANTIBODIES.COM