Bovine Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detec tion antibody, Streptavidin-HRP, recombinant bovine Interferon gamma ELISA standard, and standard reconstitution buff 1 * 576 Tests
Codice catalogo ANTIA270457-696
Nome del fornitore: ANTIBODIES.COM