Monkey Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detec tion antibody, Streptavidin-AP, recombinant human Interferon gamma ELISA standard, and standard reconstitution buffer 1 * 1.920 Tests
Catalog # ANTIA270424-2096
Supplier: ANTIBODIES.COM