Mouse IL-1 alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-AP, recombinant mouse IL-1a ELISA standard, and standard reconstitution buffer. This matched ant 1 * 1.920 Tests
Codice catalogo ANTIA270437-2096
Nome del fornitore: ANTIBODIES.COM