ContactUsLinkComponent
Hamster Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal dete ction antibody, Streptavidin-HRP, recombinant hamster Interferon gamma ELISA standard, and standard reconstitution bu 1 * 1.920 Tests
Catalog # ANTIA270372-2096
Supplier:  ANTIBODIES.COM
New Product
Hamster Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal dete ction antibody, Streptavidin-HRP, recombinant hamster Interferon gamma ELISA standard, and standard reconstitution bu 1 * 1.920 Tests
Catalog # ANTIA270372-2096
Supplier:  ANTIBODIES.COM