Human IL-5 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-AP, recombinant human IL-5 ELISA standard, and standard reconstitution buffer. This matched antibody p 1 * 576 Tests
Codice catalogo ANTIA270414-696
Nome del fornitore: ANTIBODIES.COM