ContactUsLinkComponent
Mouse pan IFN alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-AP, recombinant mouse IFN alpha ELISA standard, and standard reconstitution buffer. This matc 1 * 576 Tests
Codice catalogo ANTIA270447-696
Nome del fornitore:  ANTIBODIES.COM
New Product
Mouse pan IFN alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-AP, recombinant mouse IFN alpha ELISA standard, and standard reconstitution buffer. This matc 1 * 576 Tests
Codice catalogo ANTIA270447-696
Nome del fornitore:  ANTIBODIES.COM