ContactUsLinkComponent
Human IL-1 beta Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection ant ibody, Streptavidin-AP, recombinant human IL-1&#0223 ELISA standard, and standard reconstitution buffer. This matche 1 * 1.920 Tests
Codice catalogo ANTIA270399-2096
Nome del fornitore:  ANTIBODIES.COM
New Product
Human IL-1 beta Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection ant ibody, Streptavidin-AP, recombinant human IL-1&#0223 ELISA standard, and standard reconstitution buffer. This matche 1 * 1.920 Tests
Codice catalogo ANTIA270399-2096
Nome del fornitore:  ANTIBODIES.COM