ContactUsLinkComponent
Human IL-23 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-AP, recombinant human IL-23 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 576 Tests
  ANTIA270408-696
New Product
Human IL-23 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-AP, recombinant human IL-23 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 576 Tests
  ANTIA270408-696