ContactUsLinkComponent
Monkey IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antib ody, Streptavidin-AP, recombinant human IL-17A ELISA standard, and standard reconstitution buffer. This matched antib 1 * 1.920 Tests
Codice catalogo ANTIA270427-2096
Nome del fornitore:  ANTIBODIES.COM
New Product
Monkey IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antib ody, Streptavidin-AP, recombinant human IL-17A ELISA standard, and standard reconstitution buffer. This matched antib 1 * 1.920 Tests
Codice catalogo ANTIA270427-2096
Nome del fornitore:  ANTIBODIES.COM