ContactUsLinkComponent
Human EBI3 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-AP, recombinant human IL-27 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 576 Tests
Codice catalogo ANTIA270388-696
Nome del fornitore:  ANTIBODIES.COM
New Product
Human EBI3 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-AP, recombinant human IL-27 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 576 Tests
Codice catalogo ANTIA270388-696
Nome del fornitore:  ANTIBODIES.COM