ContactUsLinkComponent
Bovine Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detec tion antibody, Streptavidin-AP, recombinant bovine Interferon gamma ELISA standard, and standard reconstitution buffe 1 * 576 Tests
Codice catalogo ANTIA270373-696
Nome del fornitore:  ANTIBODIES.COM
New Product
Bovine Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detec tion antibody, Streptavidin-AP, recombinant bovine Interferon gamma ELISA standard, and standard reconstitution buffe 1 * 576 Tests
Codice catalogo ANTIA270373-696
Nome del fornitore:  ANTIBODIES.COM