Goat IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant bovine IL-17A ELISA standard, and standard reconstitution buffer. This matched antib 1 * 1.920 Tests
Codice catalogo ANTIA270470-2096
Nome del fornitore: ANTIBODIES.COM