Human IL-21 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant human IL-21 ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 1.920 Tests
Codice catalogo ANTIA270490-2096
Nome del fornitore: ANTIBODIES.COM