Bovine IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antib ody, Streptavidin-HRP, recombinant bovine IL-17A ELISA standard, and standard reconstitution buffer. This matched ant 1 * 576 Tests
Codice catalogo ANTIA270459-696
Nome del fornitore: ANTIBODIES.COM