ContactUsLinkComponent
Mouse IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-AP, recombinant mouse IL-17A ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 1.920 Tests
Codice catalogo ANTIA270441-2096
Nome del fornitore:  ANTIBODIES.COM
New Product
Mouse IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-AP, recombinant mouse IL-17A ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 1.920 Tests
Codice catalogo ANTIA270441-2096
Nome del fornitore:  ANTIBODIES.COM