Mouse IL-22 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant mouse IL-22 ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 1.920 Tests
Codice catalogo ANTIA270525-2096
Nome del fornitore: ANTIBODIES.COM