ContactUsLinkComponent
Rat IL-22 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-HRP, recombinant mouse IL-22 ELISA standard, and standard reconstitution buffer. This matched antibody d 1 * 576 Tests
  ANTIA270538-696
New Product
Rat IL-22 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-HRP, recombinant mouse IL-22 ELISA standard, and standard reconstitution buffer. This matched antibody d 1 * 576 Tests
  ANTIA270538-696