ContactUsLinkComponent
Human Perforin Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection anti body, Streptavidin-HRP, native human perforin ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 1.920 Tests
Codice catalogo ANTIA270502-2096
Nome del fornitore:  ANTIBODIES.COM
New Product
Human Perforin Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection anti body, Streptavidin-HRP, native human perforin ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 1.920 Tests
Codice catalogo ANTIA270502-2096
Nome del fornitore:  ANTIBODIES.COM